2,4,5-Trichloroaniline, 97%
2,4,5-Trichloroaniline, 97%
2,4,5-Trichloroaniline, 97%
Thermo Scientific Chemicals

2,4,5-Trichloroaniline, 97%

CAS: 636-30-6 | C6H4Cl3N | 196.455 g/mol
Have Questions?
Catalog NumberQuantity
B21113.30
also known as B21113-30
250 g
Catalog number B21113.30
also known as B21113-30
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.90
Save 11.25 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.90
Save 11.25 (15%)
Each
Chemical Identifiers
CAS636-30-6
IUPAC Name2,4,5-trichloroaniline
Molecular FormulaC6H4Cl3N
InChI KeyGUMCAKKKNKYFEB-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown
FormFlakes
Assay (GC)≥96.0%
Melting Point (clear melt)90.0-96.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only