Dimethyl adipate, 99%
Dimethyl adipate, 99%
Dimethyl adipate, 99%
Thermo Scientific Chemicals

Dimethyl adipate, 99%

CAS: 627-93-0 | C8H14O4 | 174.196 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number B21174.0B
also known as B21174-0B
Price (USD)
115.00
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
115.00
Each
Chemical Identifiers
CAS627-93-0
IUPAC Name1,6-dimethyl hexanedioate
Molecular FormulaC8H14O4
InChI KeyUDSFAEKRVUSQDD-UHFFFAOYSA-N
SMILESCOC(=O)CCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.4270-1.4300 @ 20?C
Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.

Solubility
Miscible with alcohols and ether. Immiscible with water.

Notes
Moisture sensitive. Incompatible with acids, bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Jastrzebski, R.; van den Berg, E. J.; Weckhuysen, B. M.; Bruijnincx, P. C. A. Sustainable production of dimethyl adipate by non-heme iron(III) catalysed oxidative cleavage of catechol. Catal. Sci. Technol. 2015, 5 (4), 2103-2109.
  2. Diogo, J. C. F.; Caetano, F. J. P.; Fareleira, J. M. N. A. Viscosity and density measurements of compressed liquid dimethyl adipate using oscillating body techniques. Fluid Phase Equilib. 2014, 367, 85-94.