D-Tagatose, 99%
D-Tagatose, 99%
D-Tagatose, 99%
Thermo Scientific Chemicals

D-Tagatose, 99%

CAS: 87-81-0 | C6H12O6 | 180.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number B21192.01
also known as B21192-01
Price (USD)
39.65
Online Exclusive
44.20
Save 4.55 (10%)
Each
Quantity:
0.25 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
44.20
Save 4.55 (10%)
Each
Chemical Identifiers
CAS87-81-0
IUPAC Name(3S,4S,5R)-2-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyLKDRXBCSQODPBY-OEXCPVAWSA-N
SMILESOCC1(O)OC[C@@H](O)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-4.5° ± 1.0° (c=1 in water)
Appearance (Color)White to pale cream
Melting Point (clear melt)128.5-134.5°C
FormCrystals or powder or crystalline powder
Loss on Drying≤0.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only