6-Fluoroveratraldehyde, 97+%
6-Fluoroveratraldehyde, 97+%
6-Fluoroveratraldehyde, 97+%
Thermo Scientific Chemicals

6-Fluoroveratraldehyde, 97+%

CAS: 71924-62-4 | C9H9FO3 | 184.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21202.03
also known as B21202-03
Price (USD)
45.65
Online Exclusive
51.10
Save 5.45 (11%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
51.10
Save 5.45 (11%)
Each
Chemical Identifiers
CAS71924-62-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormPowder or solid
Free acid (titration)<1.0%
Melting Point (clear melt)95-101?C
Assay (GC)≥97.0%
6-Fluoroveratraldehyde is used in the preparation of 6-[18F]fluoroveratraldehyde via nucleophilic aromatic substitution by [18F]fluoride. It is also used in the preparation of key metabolites of 6-fluoro-DOPA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoroveratraldehyde is used in the preparation of 6-[18F]fluoroveratraldehyde via nucleophilic aromatic substitution by [18F]fluoride. It is also used in the preparation of key metabolites of 6-fluoro-DOPA.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to air.
RUO – Research Use Only

General References:

  1. David C. Furlano.; Kenneth L. Kirk. An improved synthesis of 4-fluoroveratrole. Efficient route to 6-fluoroveratraldehyde and 6-fluoro-D,L-DOPA. J. Org. Chem. 1986, 51 (21), 4073-4075.
  2. Michael T. Clark.; Duane D. Miller. New syntheses of 2-fluoroisovanillin and 5-fluorovanillin. J. Org. Chem. 1986, 51 (21), 4072-4073.