D-Sorbose, 98%
D-Sorbose, 98%
D-Sorbose, 98%
Thermo Scientific Chemicals

D-Sorbose, 98%

CAS: 3615-56-3 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
0.5 g
Catalog number B21208.02
also known as B21208-02
Price (USD)
363.65
Online exclusive
404.00
Save 40.35
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
363.65
Online exclusive
404.00
Save 40.35
Each
Chemical Identifiers
CAS3615-56-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)≥97.5%
Melting Point (clear melt)158.0-165.0?C
Optical Rotation38.7 ± 0.1? (C=5 in water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only