D-Turanose, 98%
D-Turanose, 98%
D-Turanose, 98%
Thermo Scientific Chemicals

D-Turanose, 98%

CAS: 547-25-1 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21224.03
also known as B21224-03
Price (USD)
53.65
Online Exclusive
59.90
Save 6.25 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.90
Save 6.25 (10%)
Each
Chemical Identifiers
CAS547-25-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Optical Rotation76.0 ± 1.5? (C=4 in water)
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
FormPowder
D-Turanose is used as a carbon source by bacteria, fungi, and other organisms. It is involved in intracellular sugar signaling in plant cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Turanose is used as a carbon source by bacteria, fungi, and other organisms. It is involved in intracellular sugar signaling in plant cells.

Solubility
Soluble in water and methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container. Store at room temperature.
RUO – Research Use Only

General References:

  1. Yadu B. Tewari; Robert N. Goldberg. Thermodynamics of hydrolysis of disaccharides: Lactulose, α-d-melibiose, palatinose, d-trehalose, d-turanose and 3-o-β-d-galactopyranosyl-d-arabinose. Biophysical Chemistry.1991, 40 59-67.
  2. Adam Allerhand; David Doddrell. Carbon-13 Fourier transform nuclear magnetic resonance. VII. Study of anomeric equilibriums of ketoses in water by natural-abundance carbon-13 Fourier transform nuclear magnetic resonance. D-Fructose and D-turanose. J. Am. Chem. Soc.1971, 93 (11), 2779-2781.