2-Methylhydroquinone, 99%
2-Methylhydroquinone, 99%
2-Methylhydroquinone, 99%
Thermo Scientific Chemicals

2-Methylhydroquinone, 99%

CAS: 95-71-6 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B21241.0B
also known as B21241-0B
Price (USD)
192.65
Special offer
Online exclusive
Ends: 28-Dec-2026
227.00
Save 34.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
192.65
Special offer
Online exclusive
Ends: 28-Dec-2026
227.00
Save 34.35
Each
Chemical Identifiers
CAS95-71-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)125.0-131.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only