2,6-Dichloro-3-nitropyridine, 97%
2,6-Dichloro-3-nitropyridine, 97%
2,6-Dichloro-3-nitropyridine, 97%
Thermo Scientific Chemicals

2,6-Dichloro-3-nitropyridine, 97%

CAS: 16013-85-7 | C5H2Cl2N2O2 | 192.983 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B21255.18
also known as B21255-18
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS16013-85-7
IUPAC Name2,6-dichloro-3-nitropyridine
Molecular FormulaC5H2Cl2N2O2
InChI KeySHCWQWRTKPNTEM-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)N=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Pale yellow to yellow to orange to pale brown
FormCrystals or powder or crystalline powder or fused/lumpy solid
Melting Point (clear melt)58.0-68.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only