4-tert-Butylbenzyl alcohol, 98%
4-tert-Butylbenzyl alcohol, 98%
4-tert-Butylbenzyl alcohol, 98%
Thermo Scientific Chemicals

4-tert-Butylbenzyl alcohol, 98%

CAS: 877-65-6 | C11H16O | 164.248 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21260.22
also known as B21260-22
Price (USD)
368.65
Special offer
Online exclusive
Ends: 30-Jun-2026
433.00
Save 64.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
368.65
Special offer
Online exclusive
Ends: 30-Jun-2026
433.00
Save 64.35 (15%)
Each
Chemical Identifiers
CAS877-65-6
IUPAC Name(4-tert-butylphenyl)methanol
Molecular FormulaC11H16O
InChI KeyFVEINXLJOJPHLH-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(CO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.5160-1.5200 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
4-tert-Butylbenzyl alcohol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzyl alcohol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in alcohol and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Keep away from strong oxidizing agents, acid chlorides, acid anhydrides, strong acids.
RUO – Research Use Only
J Polleux.; M Antonietti.; M Niederberger. Ligand and solvent effects in the nonaqueous synthesis of highly ordered anisotropic tungsten oxide nanostructures. Journal of Materials Chemistry. 200616 (40), 3969-3975.