Serotonin hydrochloride, 98%
Serotonin hydrochloride, 98%
Serotonin hydrochloride, 98%
Serotonin hydrochloride, 98%
Thermo Scientific Chemicals

Serotonin hydrochloride, 98%

CAS: 153-98-0 | C10H13ClN2O | 212.677 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Catalog number B21263.06
also known as B21263-06
Price (USD)
485.65
Online Exclusive
539.00
Save 53.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
485.65
Online Exclusive
539.00
Save 53.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS153-98-0
IUPAC Namehydrogen 3-(2-aminoethyl)-1H-indol-5-ol chloride
Molecular FormulaC10H13ClN2O
InChI KeyMDIGAZPGKJFIAH-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CNC2=CC=C(O)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale grey or pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5% (Dry wt. basis, material sourced in UK)
Assay from Supplier's CofA≥97.5% (HPLC)
CommentMaterial sourced in UK or US
View more
Serotonin hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Serotonin Hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.

Solubility
Soluble in water, alcohol, hydrochloric acid and dimethyl sulfoxide. Insoluble in ether and chloroform.

Notes
Store in a cool place. Light sensitive and hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Van Schoors, J.; Lens, C.; Maes, K.; Michotte, Y.; Smolders, I.; Van Eeckhaut, A. Reassessment of the antioxidative mixture for the challenging electrochemical determination of dopamine, noradrenaline and serotonin in microdialysis samples. J. Chromatogr. B 2015, 998, 63-71.
  2. Rose, T. M.; Reilly, C. A.; Deering-Rice, C. E.; Brewster, C.; Brewster, C. Inhibition of FAAH, TRPV1, and COX2 by NSAID-serotonin conjugates. Bioorg. Med. Chem. Lett. 2014, 24 (24), 5695-5698.