3-Carboxybenzaldehyde, 97%
3-Carboxybenzaldehyde, 97%
3-Carboxybenzaldehyde, 97%
Thermo Scientific Chemicals

3-Carboxybenzaldehyde, 97%

CAS: 619-21-6 | C8H6O3 | 150.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21277.03
also known as B21277-03
Price (USD)
62.65
Online Exclusive
69.70
Save 7.05 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
62.65
Online Exclusive
69.70
Save 7.05 (10%)
Each
Chemical Identifiers
CAS619-21-6
IUPAC Name3-formylbenzoic acid
Molecular FormulaC8H6O3
InChI KeyUHDNUPHSDMOGCR-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
3-Formylbenzoic acid is used in the synthesis of bicyclic cis-2-azetidinone derivatives via Ugi 4-centre 3-component reaction, porphyrin capped with a steroidal superstructure bearing convergent hydroxy groups and 3-[(4-amino-1,2-dihydro-1-oxo-2-phenyl-1,2,4-triazolo[4,3-a]quinoxalin-6-yl)amino]methyl benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Formylbenzoic acid is used in the synthesis of bicyclic cis-2-azetidinone derivatives via Ugi 4-centre 3-component reaction, porphyrin capped with a steroidal superstructure bearing convergent hydroxy groups and 3-[(4-amino-1,2-dihydro-1-oxo-2-phenyl-1,2,4-triazolo[4,3-a]quinoxalin-6-yl)amino]methyl benzoic acid.

Solubility
Soluble in methanol (100 mg/mL) gives a clear to slightly hazy, colorless to very faintly brown.

Notes
Air sensitive. Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Gedey S, et al. Synthesis of Alicyclic-lactams via the Ugi Reaction on a Solid Support.Lett. Org. Chem.20041(3), 215-20.
  2. Richard P and Jeremy KM. Synthesis, binding properties and self-functionalization of a steroid-capped porphyrin.J. Chem. Soc. Chem. Commun.19918574-77.