tert-Butyldimethylchlorosilane, 50% w/w in toluene, Thermo Scientific Chemicals
tert-Butyldimethylchlorosilane, 50% w/w in toluene, Thermo Scientific Chemicals
tert-Butyldimethylchlorosilane, 50% w/w in toluene, Thermo Scientific Chemicals
Thermo Scientific™

tert-Butyldimethylchlorosilane, 50% w/w in toluene, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B21286.18
also known as B21286-18
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B21286.18
also known as B21286-18
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS18162-48-6
IUPAC Nametert-butyl(chloro)dimethylsilane
Molecular FormulaC6H15ClSi
InChI KeyBCNZYOJHNLTNEZ-UHFFFAOYSA-N
SMILESCC(C)(C)[Si](C)(C)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (Titration ex Chloride)48-52%
Assay (GC)≥97.0% (dry wt)
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only