Methyl 2-bromo-5-nitrobenzoate, 98+%
Methyl 2-bromo-5-nitrobenzoate, 98+%
Methyl 2-bromo-5-nitrobenzoate, 98+%
Methyl 2-bromo-5-nitrobenzoate, 98+%
Thermo Scientific Chemicals

Methyl 2-bromo-5-nitrobenzoate, 98+%

CAS: 6942-36-5 | C8H6BrNO4 | 260.043 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21288.14
also known as B21288-14
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS6942-36-5
IUPAC Namemethyl 2-bromo-5-nitrobenzoate
Molecular FormulaC8H6BrNO4
InChI KeyVSEYYEKRZNRECT-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=CC=C1Br)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point (clear melt)78.0-84.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only