2,6-Dichloropurine, 97%
2,6-Dichloropurine, 97%
2,6-Dichloropurine, 97%
Thermo Scientific Chemicals

2,6-Dichloropurine, 97%

CAS: 5451-40-1 | C5H2Cl2N4 | 189.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21289.06
also known as B21289-06
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS5451-40-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or pale pink
FormPowder
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only