Diisopropyl sulfide, 99%
Diisopropyl sulfide, 99%
Diisopropyl sulfide, 99%
Thermo Scientific Chemicals

Diisopropyl sulfide, 99%

CAS: 625-80-9 | C6H14S | 118.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B21302.18
also known as B21302-18
Price (USD)
132.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
132.00
Each
Chemical Identifiers
CAS625-80-9
IUPAC Name2-(propan-2-ylsulfanyl)propane
Molecular FormulaC6H14S
InChI KeyXYWDPYKBIRQXQS-UHFFFAOYSA-N
SMILESCC(C)SC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4370-1.4400 @ 20?C
FormLiquid
Used as a flavoring agents and intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agents and intermediate for organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Charles W. Rees, Oleg A. Rakitin, Carlos F. Marcos, Tomás Torroba . Synthesis of Sulfur-Rich 1,2- and 1,3-Dithiolo Disulfides and Thiodesaurines from Diisopropyl Sulfide. J. Org. Chem., 1999, 64(12), 4376-4380.
  2. D. T. McAllan; T. V. Cullum; R. A. Dean; F. A. Fidler. The Preparation and Properties of Sulfur Compounds Related to Petroleum. I. The Dialkyl Sulfides and Disulfides. J. Am. Chem. Soc., 1951, 73(8), 3627-3632.