Calix[4]arene, 98%
Calix[4]arene, 98%
Calix[4]arene, 98%
Thermo Scientific Chemicals

Calix[4]arene, 98%

CAS: 74568-07-3 | C28H24O4 | 424.496 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21320.06
also known as B21320-06
Price (USD)
329.65
Online Exclusive
366.00
Save 36.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
329.65
Online Exclusive
366.00
Save 36.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS74568-07-3
IUPAC Namepentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
Molecular FormulaC28H24O4
InChI KeyYPNHVQZZPXPQOS-UHFFFAOYSA-N
SMILESOC1=C2CC3=CC=CC(CC4=CC=CC(CC5=CC=CC(CC1=CC=C2)=C5O)=C4O)=C3O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to cream
Assay (HPLC)≥97.5%
Functionalized calix[4]arenes can bind DNA and induce cell transfection. Calixarenes lend themselves well to many applications because of the multiplicity of options for such structural elaboration such as in electrochemical sensors, optical sensors, chiral recognition devices,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Functionalized calix[4]arenes can bind DNA and induce cell transfection. Calixarenes lend themselves well to many applications because of the multiplicity of options for such structural elaboration such as in electrochemical sensors, optical sensors, chiral recognition devices,

Solubility
Soluble in chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Kenny B. Lipkowitz and Greg Pearl. Structural features of solid-state calix[4]arene in the cone conformation.J. Org. Chem.,199358(24), 6729-6736.
  2. Sergio Sanz, et al. Calix[4]arene-supported rare earth octahedral.Chem. Commun., 201248(10), 1449-1451.