2-Nitrobenzhydrazide, 98%
2-Nitrobenzhydrazide, 98%
2-Nitrobenzhydrazide, 98%
Thermo Scientific Chemicals

2-Nitrobenzhydrazide, 98%

CAS: 606-26-8 | C7H7N3O3 | 181.151 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21330.06
also known as B21330-06
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
42.50
Save 5.85
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
42.50
Save 5.85
Each
Chemical Identifiers
CAS606-26-8
IUPAC Name2-nitrobenzohydrazide
Molecular FormulaC7H7N3O3
InChI KeyLYGGDXLOJMNFBV-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to brown
FormPowder or crystalline powder
Assay (HPLC)≥97.5%
Melting Point (clear melt)118.0-125.0?C
The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.

Solubility
Soluble in methanol(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. H. M. Ali,; S. N. Abdul Halim,; S. W. Ng. Bis[5-bromo-1H-indole-3-carbaldehyde (2-nitro­benzo­yl)hydrazonato-[kappa]2N,O]bis­(pyridine-[kappa]N)nickel(II) pyridine disolvate. Acta Cryst. 2005, E61, m1682-m1683.
  2. F. I. Carrolla,; H. Wayne Millera. The Reaction Of 2,3-Dichloro-1,4-Naphthoquinone With P-Nitrobenzhydrazide1 . Organic Preparations and Procedures. 1970, 2 (4), 259-263.