2-Amino-5-nitrothiazole, 97%
2-Amino-5-nitrothiazole, 97%
2-Amino-5-nitrothiazole, 97%
2-Amino-5-nitrothiazole, 97%
Thermo Scientific Chemicals

2-Amino-5-nitrothiazole, 97%

CAS: 121-66-4 | C3H3N3O2S | 145.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B21335.22
also known as B21335-22
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.70
Save 13.05 (14%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.70
Save 13.05 (14%)
Each
Add to cart
Chemical Identifiers
CAS121-66-4
IUPAC Name5-nitro-1,3-thiazol-2-amine
Molecular FormulaC3H3N3O2S
InChI KeyMIHADVKEHAFNPG-UHFFFAOYSA-N
SMILESNC1=NC=C(S1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only