2,5-Dichloro-p-xylene, 98%
2,5-Dichloro-p-xylene, 98%
2,5-Dichloro-p-xylene, 98%
Thermo Scientific Chemicals

2,5-Dichloro-p-xylene, 98%

CAS: 1124-05-6 | C8H8Cl2 | 175.052 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B21344.22
also known as B21344-22
Price (USD)
167.65
Online Exclusive
186.00
Save 18.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
167.65
Online Exclusive
186.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS1124-05-6
IUPAC Name1,4-dichloro-2,5-dimethylbenzene
Molecular FormulaC8H8Cl2
InChI KeyUTGSRNVBAFCOEU-UHFFFAOYSA-N
SMILESCC1=CC(Cl)=C(C)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)66.0-72.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only