3,5-Difluorobenzonitrile, 99%
3,5-Difluorobenzonitrile, 99%
3,5-Difluorobenzonitrile, 99%
3,5-Difluorobenzonitrile, 99%
Thermo Scientific Chemicals

3,5-Difluorobenzonitrile, 99%

CAS: 64248-63-1 | C7H3F2N | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21347.14
also known as B21347-14
Price (USD)
134.65
Online Exclusive
149.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
134.65
Online Exclusive
149.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS64248-63-1
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)84.0-91.0?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only