Valeraldehyde, 96%
Valeraldehyde, 96%
Valeraldehyde, 96%
Thermo Scientific Chemicals

Valeraldehyde, 96%

CAS: 110-62-3 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B21389.0B
also known as B21389-0B
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS110-62-3
IUPAC Namepentanal
Molecular FormulaC5H10O
InChI KeyHGBOYTHUEUWSSQ-UHFFFAOYSA-N
SMILESCCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥95.0%
Free acid (titration)≤1.5%
Refractive Index1.3905-1.3970 @ 20°C
It finds it application in flavorings, resin chemistry, and rubber accelerators. Valeraldehyde is used as a gaseous standard in the study of the sorptive loss pattern for volatile compounds. Valeraldehyde is used as an intermediate in industrial settings in the production of valeric acid and amyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application in flavorings, resin chemistry, and rubber accelerators. Valeraldehyde is used as a gaseous standard in the study of the sorptive loss pattern for volatile compounds. Valeraldehyde is used as an intermediate in industrial settings in the production of valeric acid and amyl alcohol.

Solubility
Soluble in water at 20°C, 13.5g/L. Soluble in alcohol.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Wang Y, et.al. Evaluation of n-valeraldehyde modified chitosan as a matrix for hydrophobic interaction chromatography.J Chromatogr. A.2002952(1-2), 79-83.
  2. Tsai SW, et al. Time-weighted average sampling of airborne n-valeraldehyde by a solid-phase microextration device.J Chromatogr. A., 2002954(1-2), 191-8.