3,5-Difluorobenzoyl chloride, 98+%
3,5-Difluorobenzoyl chloride, 98+%
3,5-Difluorobenzoyl chloride, 98+%
3,5-Difluorobenzoyl chloride, 98+%
Thermo Scientific Chemicals

3,5-Difluorobenzoyl chloride, 98+%

CAS: 129714-97-2 | C7H3ClF2O | 176.547 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21400.06
also known as B21400-06
Price (USD)
64.65
Online Exclusive
71.20
Save 6.55 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
64.65
Online Exclusive
71.20
Save 6.55 (9%)
Each
Add to cart
Chemical Identifiers
CAS129714-97-2
IUPAC Name3,5-difluorobenzoyl chloride
Molecular FormulaC7H3ClF2O
InChI KeyOYZWEOORLJBPMA-UHFFFAOYSA-N
SMILESFC1=CC(=CC(F)=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.0%
Identification (FTIR)Conforms
Refractive Index1.5015-1.5055 @ 20°C
3,5-Difluorobenzoyl chloride, is used as an pharmaceutical intermediate, organic intermediate and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluorobenzoyl chloride, is used as an pharmaceutical intermediate, organic intermediate and chemical synthesis.

Solubility
It reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids and bases. Moisture Sensitive
RUO – Research Use Only

General References:

  1. F Chu.; CJ Hawker. A versatile synthesis of isomeric hyperbranched polyetherketones. Polymer Bulletin. 199330 (1), 265-272.
  2. MB Andrus.; J Liu. Synthesis of polyhydroxylated ester analogs of the stilbene resveratrol using decarbonylative Heck couplings. Tetrahedron letters. 200647 (32), 5811-5814.