1,1-Diphenyl-2-propyn-1-ol, 99%
1,1-Diphenyl-2-propyn-1-ol, 99%
1,1-Diphenyl-2-propyn-1-ol, 99%
1,1-Diphenyl-2-propyn-1-ol, 99%
Thermo Scientific Chemicals

1,1-Diphenyl-2-propyn-1-ol, 99%

CAS: 3923-52-2 | C15H12O | 208.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21428.06
also known as B21428-06
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.70
Save 7.05 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.70
Save 7.05 (15%)
Each
Add to cart
Chemical Identifiers
CAS3923-52-2
IUPAC Name1,1-diphenylprop-2-yn-1-ol
Molecular FormulaC15H12O
InChI KeySMCLTAARQYTXLW-UHFFFAOYSA-N
SMILESOC(C#C)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)45.0-52.0?C
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder or fused solid
1,1-Diphenyl-2-propyn-1-ol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Diphenyl-2-propyn-1-ol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in medicine.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. P Crochet.; MA Esteruelas.; E Gutiérrez-Puebla. Unusual Activation of 1, 1-Diphenyl-2-propyn-1-ol Mediated by the Os (η5-C5H5) Unit. Organometallics. 199817 (15), 3141-3142.
  2. E Negishi.; M Hata.; C Xu. A Strictly Pair-Selective Synthesis of Conjugated Diynes via Pd-Catalyzed Cross Coupling of 1,3-Diynylzincs:  A Superior Alternative to the Cadiot-Chodkiewicz Reaction. Organic letters. 20002 (23), 3687-3689.