2,3,6-Trimethylquinoxaline, 97%
2,3,6-Trimethylquinoxaline, 97%
2,3,6-Trimethylquinoxaline, 97%
Thermo Scientific Chemicals

2,3,6-Trimethylquinoxaline, 97%

CAS: 17635-21-1 | C11H12N2 | 172.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21432.06
also known as B21432-06
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS17635-21-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to brown
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)88.5-97.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only