4-tert-Butylcalix[8]arene, 96%
4-tert-Butylcalix[8]arene, 96%
4-tert-Butylcalix[8]arene, 96%
Thermo Scientific Chemicals

4-tert-Butylcalix[8]arene, 96%

CAS: 68971-82-4 | C88H112O8 | 1297.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21443.14
also known as B21443-14
Price (USD)
448.65
Online Exclusive
498.00
Save 49.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
448.65
Online Exclusive
498.00
Save 49.35 (10%)
Each
Chemical Identifiers
CAS68971-82-4
IUPAC Name5,11,17,23,29,35,41,47-octa-tert-butylnonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
Molecular FormulaC88H112O8
InChI KeyOLZFZIXORGGLLS-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC8=C(O)C(CC9=C(O)C(C2)=CC(=C9)C(C)(C)C)=CC(=C8)C(C)(C)C)=CC(=C7)C(C)(C)C)=CC(=C6)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥95.0%
Purity (DSC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only