4-Bromo-3,5-dimethoxybenzoic acid, 98%
4-Bromo-3,5-dimethoxybenzoic acid, 98%
4-Bromo-3,5-dimethoxybenzoic acid, 98%
4-Bromo-3,5-dimethoxybenzoic acid, 98%
Thermo Scientific Chemicals

4-Bromo-3,5-dimethoxybenzoic acid, 98%

CAS: 56518-42-4 | C9H8BrO4 | 260.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21449.06
also known as B21449-06
Price (USD)
42.65
Online Exclusive
47.60
Save 4.95 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
42.65
Online Exclusive
47.60
Save 4.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS56518-42-4
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)253-259?C
Appearance (Color)White to pale cream
FormCrystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only