alpha-Bisabolol, 96%
alpha-Bisabolol, 96%
alpha-Bisabolol, 96%
Thermo Scientific Chemicals

alpha-Bisabolol, 96%

CAS: 515-69-5 | C15H26O | 222.372 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B21457.18
also known as B21457-18
Price (USD)
212.65
Online Exclusive
236.00
Save 23.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
212.65
Online Exclusive
236.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS515-69-5
IUPAC Name(2S)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
Molecular FormulaC15H26O
InChI KeyRGZSQWQPBWRIAQ-GJZGRUSLSA-N
SMILESCC(C)=CCC[C@](C)(O)[C@@H]1CCC(C)=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4925-1.4985 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid or viscous liquid
Assay (GC)≥95.0%
It finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.

Solubility
Soluble in alcohol, natural, mineral and synthetic oils. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Forrer M, et.al. The antimicrobial activity of alpha-bisabolol and tea tree oil against Solobacterium moorei, a Gram-positive bacterium associated with halitosis.Arch Oral Biol.201358(1), 10-6.
  2. Costarelli L, et al. In vivo effect of alpha-bisabolol, a nontoxic sesquiterpene alcohol, on the induction of spontaneous mammary tumors in HER-2eu transgenic mice.Oncol Res., 201018(9), 409-18.