2-Amino-5-fluorobenzoic acid, 97%
2-Amino-5-fluorobenzoic acid, 97%
2-Amino-5-fluorobenzoic acid, 97%
Thermo Scientific Chemicals

2-Amino-5-fluorobenzoic acid, 97%

CAS: 446-08-2 | C7H6FNO2 | 155.128 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21458.14
also known as B21458-14
Price (USD)
749.65
Online Exclusive
833.00
Save 83.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
749.65
Online Exclusive
833.00
Save 83.35 (10%)
Each
Chemical Identifiers
CAS446-08-2
IUPAC Name2-amino-5-fluorobenzoic acid
Molecular FormulaC7H6FNO2
InChI KeyFPQMGQZTBWIHDN-UHFFFAOYSA-N
SMILESNC1=CC=C(F)C=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0%
Assay (Silylated GC)≥96.0%
Appearance (Color)Pale cream to cream to brown or yellow
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only