4,4,4-Trifluoro-1-butanol, 97%
4,4,4-Trifluoro-1-butanol, 97%
4,4,4-Trifluoro-1-butanol, 97%
Thermo Scientific Chemicals

4,4,4-Trifluoro-1-butanol, 97%

CAS: 461-18-7 | C4H7F3O | 128.094 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21460.06
also known as B21460-06
Price (USD)
223.65
Online Exclusive
248.00
Save 24.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
223.65
Online Exclusive
248.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS461-18-7
IUPAC Name4,4,4-trifluorobutan-1-ol
Molecular FormulaC4H7F3O
InChI KeyVKRFUGHXKNNIJO-UHFFFAOYSA-N
SMILESOCCCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.3405-1.3455 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only