2,6-Dihydroxynaphthalene, 98%
2,6-Dihydroxynaphthalene, 98%
2,6-Dihydroxynaphthalene, 98%
Thermo Scientific Chemicals

2,6-Dihydroxynaphthalene, 98%

CAS: 581-43-1 | C10H8O2 | 160.172 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21465.06
also known as B21465-06
Price (USD)
69.65
Online Exclusive
77.10
Save 7.45 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
69.65
Online Exclusive
77.10
Save 7.45 (10%)
Each
Chemical Identifiers
CAS581-43-1
IUPAC Namenaphthalene-2,6-diol
Molecular FormulaC10H8O2
InChI KeyMNZMMCVIXORAQL-UHFFFAOYSA-N
SMILESOC1=CC2=CC=C(O)C=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)220.0-227.0?C
Appearance (Color)Pale cream to cream to pale brown to pale gray to gray
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
2,6-Dihydroxynaphthalene is used in the preparation of 1,5-dichloro-2,6-diethynylnaphthalenes. It is used to produce convulsions in mice. It exhibits antiproliferative activity towards certain cells. It is also employed in the preparation of the first-generation rotaxane dendrimer. Further, it is used in the production of several dyes, pigments, fluorescent whiteners, tanning agents, antioxidants and antiseptics. In addition to this, it is converted into amines, esters, ethers and carboxylic derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dihydroxynaphthalene is used in the preparation of 1,5-dichloro-2,6-diethynylnaphthalenes. It is used to produce convulsions in mice. It exhibits antiproliferative activity towards certain cells. It is also employed in the preparation of the first-generation rotaxane dendrimer. Further, it is used in the production of several dyes, pigments, fluorescent whiteners, tanning agents, antioxidants and antiseptics. In addition to this, it is converted into amines, esters, ethers and carboxylic derivatives.

Solubility
Soluble in methanol and hot water. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Shankar, K.; Kirillov, A. M.; Baruah, J. B. A modular approach for molecular recognition by zinc dipicolinate complexes. Dalton Trans. 2015, 44 (32), 14411-14423.
  2. Zhou, T. Y.; Qi, Q. Y.; Zhang, Y.; Xu, X. N.; Zhao, X. A thermally stable pH-responsive supramolecular buckle based on the encapsulation of 4-(4-aminophenyl)-N-methylpyridinium by cucurbit[8]uril. Org. Chem. Front. 2015, 2 (9), 1030-1034.