2,5-Dimethylstyrene, 97%, stab. with 4-tert-butylcatechol
2,5-Dimethylstyrene, 97%, stab. with 4-tert-butylcatechol
2,5-Dimethylstyrene, 97%, stab. with 4-tert-butylcatechol
Thermo Scientific Chemicals

2,5-Dimethylstyrene, 97%, stab. with 4-tert-butylcatechol

CAS: 2039-89-6 | C10H12 | 132.206 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21476.03
also known as B21476-03
Price (USD)
30.65
Online Exclusive
34.20
Save 3.55 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
34.20
Save 3.55 (10%)
Each
Chemical Identifiers
CAS2039-89-6
IUPAC Name2-ethenyl-1,4-dimethylbenzene
Molecular FormulaC10H12
InChI KeyDBWWINQJTZYDFK-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C(C=C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colourless to pale yellow to pale orange
Assay (GC)≥96.0%
Refractive Index1.5365-1.5415 @20?C
Solution Test10% in diethyl ether: clear
Solution Test10% in methanol: clear
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only