2,5-Dichloro-p-phenylenediamine, 99%
2,5-Dichloro-p-phenylenediamine, 99%
2,5-Dichloro-p-phenylenediamine, 99%
2,5-Dichloro-p-phenylenediamine, 99%
Thermo Scientific Chemicals

2,5-Dichloro-p-phenylenediamine, 99%

CAS: 20103-09-7 | C6H6Cl2N2 | 177.028 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21494.06
also known as B21494-06
Price (USD)
39.20
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.20
Each
Add to cart
Chemical Identifiers
CAS20103-09-7
IUPAC Name2,5-dichlorobenzene-1,4-diamine
Molecular FormulaC6H6Cl2N2
InChI KeyQAYVHDDEMLNVMO-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=C(N)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to brown
FormCrystals or powder or crystalline flakes/powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (GC)≥98.5%
Melting Point (clear melt)160-167°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only