2-Chloromandelic acid, 98%
2-Chloromandelic acid, 98%
2-Chloromandelic acid, 98%
Thermo Scientific Chemicals

2-Chloromandelic acid, 98%

CAS: 10421-85-9 | C8H7ClO3 | 186.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21525.14
also known as B21525-14
Price (USD)
101.65
Special offer
Online exclusive
Ends: 30-Jun-2026
119.00
Save 17.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
101.65
Special offer
Online exclusive
Ends: 30-Jun-2026
119.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS10421-85-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
2-Chloromandelic acid, is used as a pharmaceutical intermediate as well as in the medicine industry. It is used as a intermediate of clopidrogrel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloromandelic acid, is used as a pharmaceutical intermediate as well as in the medicine industry. It is used as a intermediate of clopidrogrel.

Solubility
Very soluble in water.

Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Q He.; S Rohani.; J Zhu.; H Gomaa. Crystallization of the racemic compound and conglomerate of (RS)-2-chloromandelic acid. Crystal Growth & Design. 201010 (12), 5136-5145.
  2. Y Peng.; Q He.; S Rohani, H Jenkins. Resolution of 2-chloromandelic acid with (R)-(+)-N-benzyl-1-phenylethylamine: chiral discrimination mechanism. Chirality. 201224 (5), 349-355.