Xylenecyanol FF, dye content 70%
Xylenecyanol FF, dye content 70%
Xylenecyanol FF, dye content 70%
Thermo Scientific Chemicals

Xylenecyanol FF, dye content 70%

CAS: 2650-17-1 | C25H27N2NaO6S2 | 538.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B21530.09
also known as B21530-09
Price (USD)
54.30
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
54.30
Each
Chemical Identifiers
CAS2650-17-1
IUPAC Namesodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
Molecular FormulaC25H27N2NaO6S2
InChI KeyVVLFAAMTGMGYBS-KRQUPCAFSA-M
SMILES[Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to red / brown
FormPowder
Assay (Dye)≥70%
CommentSuitable as a tracking dye for DNA sequencing.
Loss on Drying<8.0% (105?C / constant wt.)
View more
Xylene Cyanol FF is a tracking dye commonly used for nucleic acids. The compound has been used in both electrophoresis polyacrylamide and agarose gels for various evaluations including DNA sequencing. The compound has also been used in a mobility shift analysis of ssDNA. In addition, two highly sensitive spectrophotometric assays have been developed for the detection of platinum (IV) (sc-250738, Platinum(IV) chloride) and Chromium (sc-214714) with the use of Xylene Cyanol FF. As an organic dye-inactivated Sendai virus vaccine, Xylene Cyanol FF displayed strong protection against the virus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xylene Cyanol FF is a tracking dye commonly used for nucleic acids. The compound has been used in both electrophoresis polyacrylamide and agarose gels for various evaluations including DNA sequencing. The compound has also been used in a mobility shift analysis of ssDNA. In addition, two highly sensitive spectrophotometric assays have been developed for the detection of platinum (IV) (sc-250738, Platinum(IV) chloride) and Chromium (sc-214714) with the use of Xylene Cyanol FF. As an organic dye-inactivated Sendai virus vaccine, Xylene Cyanol FF displayed strong protection against the virus.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
RUO – Research Use Only
HD Revanasiddappa.; TNK Kumar. A highly sensitive spectrophotometric determination of chromium using leuco Xylene cyanol FF. Talanta. 200360 (1), 1-8.