2-Fluoro-4-hydroxybenzonitrile, 99%
2-Fluoro-4-hydroxybenzonitrile, 99%
2-Fluoro-4-hydroxybenzonitrile, 99%
Thermo Scientific Chemicals

2-Fluoro-4-hydroxybenzonitrile, 99%

CAS: 82380-18-5 | C7H4FNO | 137.113 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21535.06
also known as B21535-06
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS82380-18-5
IUPAC Name2-fluoro-4-hydroxybenzonitrile
Molecular FormulaC7H4FNO
InChI KeyREIVHYDACHXPNH-UHFFFAOYSA-N
SMILESOC1=CC=C(C#N)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)121.0-125.0?C
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only