2-Chlorostyrene, 95%, stab. with 0.1% 4-tert-butylcatechol
2-Chlorostyrene, 95%, stab. with 0.1% 4-tert-butylcatechol
2-Chlorostyrene, 95%, stab. with 0.1% 4-tert-butylcatechol
Thermo Scientific Chemicals

2-Chlorostyrene, 95%, stab. with 0.1% 4-tert-butylcatechol

CAS: 2039-87-4 | C8H7Cl | 138.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21551.14
also known as B21551-14
Price (USD)
279.65
Online Exclusive
311.00
Save 31.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
279.65
Online Exclusive
311.00
Save 31.35 (10%)
Each
Chemical Identifiers
CAS2039-87-4
IUPAC Name1-chloro-2-ethenylbenzene
Molecular FormulaC8H7Cl
InChI KeyISRGONDNXBCDBM-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Refractive Index1.5625-1.5675 @ 20°C
Solution Test10% in diethyl ether: clear
Solution Test10% in methanol: clear
View more
2-Chlorostyrene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorostyrene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only
M Okada.; K Fujimoto.; T Nose. Phase separation induced by polymerization of 2-chlorostyrene in a polystyrene/dibutyl phthalate mixture. Macromolecules. 199528 (6), 1795-1800.