Dimethyl 5-hydroxyisophthalate, 98%
Dimethyl 5-hydroxyisophthalate, 98%
Dimethyl 5-hydroxyisophthalate, 98%
Dimethyl 5-hydroxyisophthalate, 98%
Thermo Scientific Chemicals

Dimethyl 5-hydroxyisophthalate, 98%

CAS: 13036-02-7 | C10H10O5 | 210.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B21565.18
also known as B21565-18
Price (USD)
109.65
Online Exclusive
122.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
109.65
Online Exclusive
122.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS13036-02-7
IUPAC Name1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate
Molecular FormulaC10H10O5
InChI KeyDOSDTCPDBPRFHQ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=CC(O)=C1)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)160.0-167.0?C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Assay (HPLC)≥97.5%
Dimethyl 5-Hydroxyisophthalate is a useful synthetic intermediate in the synthesis of N,N’-Bis(2,3-dihydroxypropyl)-5-[(N-(2-hydroxyethyl)carbamoyl)methoxy]-2,4,6-triiodoisophthalamide (B426310); a compound related to Ioversol( I737000) which is a nonionic, low osmolality, radiographic contrast agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-Hydroxyisophthalate is a useful synthetic intermediate in the synthesis of N,N’-Bis(2,3-dihydroxypropyl)-5-[(N-(2-hydroxyethyl)carbamoyl)methoxy]-2,4,6-triiodoisophthalamide (B426310); a compound related to Ioversol( I737000) which is a nonionic, low osmolality, radiographic contrast agent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

General References:

  1. D Parker.; WJ Feast. Synthesis, structure, and properties of hyperbranched polyesters based on dimethyl 5-(2-hydroxyethoxy) isophthalate. Macromolecules. 200134 (7), 2048-2059.
  2. WJ Feast.; NM Stainton. Synthesis, structure and properties of some hyperbranched polyesters. Journal of Materials Chemistry. 19955 (3), 405-411.