Thermo Scientific Chemicals

Perfluoropentanoic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21567.14
25 g, Each
Thermo Scientific Chemicals

Perfluoropentanoic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21567.14
25 g, Each
Quantity
Catalog number: B21567.14
also known as B21567-14
Price (USD)
Price: 245.00
Online price: 208.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2706-90-3
Form
Liquid
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.2925-1.3015 @ 20?C

Description

It is used in the analysis of perfluorooctanesulfonate and related fluorochemicals in water and biological tissue samples by liquid chromatography-ion trap mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the analysis of perfluorooctanesulfonate and related fluorochemicals in water and biological tissue samples by liquid chromatography-ion trap mass spectrometry.

Solubility
Partly soluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text