4-Ethylresorcinol, 98%
4-Ethylresorcinol, 98%
4-Ethylresorcinol, 98%
4-Ethylresorcinol, 98%
Thermo Scientific Chemicals

4-Ethylresorcinol, 98%

CAS: 2896-60-8 | C8H10O2 | 138.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21575.06
also known as B21575-06
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.40
Save 9.75 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.40
Save 9.75 (15%)
Each
Add to cart
Chemical Identifiers
CAS2896-60-8
IUPAC Name4-ethylbenzene-1,3-diol
Molecular FormulaC8H10O2
InChI KeyVGMJYYDKPUPTID-UHFFFAOYSA-N
SMILESCCC1=CC=C(O)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder
Melting Point (clear melt)94.0-100.0?C
It is employed as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Solubility in methanol; almost transparency. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Jacob Ehrlich. The Reduction of p-Hydroxyformanilide by Lithium Aluminum Hydride to N-Methyl-p-aminophenol.J. Am. Chem. Soc.,194870(6),2286-2287.
  2. R.W. Thring and E. Chornet. Analysis of phenols from lignin depolymerization by capillary gas chromatography.J. Chromatogr.. A.,1989467441-446.