1-Bromo-2,3-difluorobenzene, 98%
1-Bromo-2,3-difluorobenzene, 98%
1-Bromo-2,3-difluorobenzene, 98%
Thermo Scientific Chemicals

1-Bromo-2,3-difluorobenzene, 98%

CAS: 38573-88-5 | C6H3BrF2 | 192.991 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21585.03
also known as B21585-03
Price (USD)
46.65
Online Exclusive
52.20
Save 5.55 (11%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
46.65
Online Exclusive
52.20
Save 5.55 (11%)
Each
Chemical Identifiers
CAS38573-88-5
IUPAC Name1-bromo-2,3-difluorobenzene
Molecular FormulaC6H3BrF2
InChI KeyRKWWASUTWAFKHA-UHFFFAOYSA-N
SMILESFC1=CC=CC(Br)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to yellow
FormLiquid
Refractive Index1.5080-1.5120 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only