2'-Acetoxyacetophenone, 98%
2'-Acetoxyacetophenone, 98%
2'-Acetoxyacetophenone, 98%
Thermo Scientific Chemicals

2'-Acetoxyacetophenone, 98%

CAS: 7250-94-4 | C10H10O3 | 178.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21624.06
also known as B21624-06
Price (USD)
29.65
Online Exclusive
33.00
Save 3.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
29.65
Online Exclusive
33.00
Save 3.35 (10%)
Each
Chemical Identifiers
CAS7250-94-4
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)85-92?C
Appearance (Color)White to cream
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only