Methyl 4-acetylbenzoate, 99%
Methyl 4-acetylbenzoate, 99%
Methyl 4-acetylbenzoate, 99%
Thermo Scientific Chemicals

Methyl 4-acetylbenzoate, 99%

CAS: 3609-53-8 | C10H10O3 | 178.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number B21626.04
also known as B21626-04
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
71.90
Save 10.25
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
71.90
Save 10.25
Each
Chemical Identifiers
CAS3609-53-8
IUPAC Namemethyl 4-acetylbenzoate
Molecular FormulaC10H10O3
InChI KeyQNTSFZXGLAHYLC-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)91.0-97.0?C
Assay (GC)≥98.5%
Appearance (Color)Pale cream to pale yellow to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only