4-Chloro-3-nitrobenzenesulfonyl chloride, 98%
4-Chloro-3-nitrobenzenesulfonyl chloride, 98%
4-Chloro-3-nitrobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

4-Chloro-3-nitrobenzenesulfonyl chloride, 98%

CAS: 97-08-5 | C6H3Cl2NO4S | 256.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21632.14
also known as B21632-14
Price (USD)
101.65
Online exclusive
113.00
Save 11.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
101.65
Online exclusive
113.00
Save 11.35
Each
Chemical Identifiers
CAS97-08-5
IUPAC Name4-chloro-3-nitrobenzene-1-sulfonyl chloride
Molecular FormulaC6H3Cl2NO4S
InChI KeySEWNAJIUKSTYOP-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=CC=C1Cl)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)57.0-63.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only