10-Methylphenothiazine, 98%
10-Methylphenothiazine, 98%
10-Methylphenothiazine, 98%
Thermo Scientific Chemicals

10-Methylphenothiazine, 98%

CAS: 1207-72-3 | C13H11NS | 213.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21648.14
also known as B21648-14
Price (USD)
162.65
Online Exclusive
180.00
Save 17.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
162.65
Online Exclusive
180.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS1207-72-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow or pale pink
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)96.0-104.0°C
They are used in pharmaceutical manufacturing especially antipsychotic drugs. Also used as a stabilizer against polymerization of acrylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are used in pharmaceutical manufacturing especially antipsychotic drugs. Also used as a stabilizer against polymerization of acrylates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Gregory L. McIntire; Henry N. Blount. Electrochemistry in ordered systems. 1. Oxidative electrochemistry of 10-methylphenothiazine in anionic, cationic, and nonionic micellar systems. J. Am. Chem. Soc. 1979, 101 (26), 7720-7721.
  2. Minh C. Ko; Gerald J. Meyer. Dynamic Quenching of Porous Silicon Photoluminescence by Anthracene and 10-Methylphenothiazine. Chem. Mater., 1995 , 7(1), 12-14.