3-(2-Furyl)acrylic acid is a reactant used in the preparation of heteroaryl-substituted bis-trifluoromethyl carbinols and trifluoroacetophenone derivatives malonyl-CoA decarboxylase (MCD) inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(2-Furyl)acrylic acid is a reactant used in the preparation of heteroaryl-substituted bis-trifluoromethyl carbinols and trifluoroacetophenone derivatives malonyl-CoA decarboxylase (MCD) inhibitors.
Solubility
Soluble in dichloromethane and diethyl ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Wallace, D. et al. Novel trifluoroacetophenone derivatives as malonyl-CoA decarboxylase inhibitors. Bioorganic & Medicinal Chemistry Letters. 2007, 17 (4), 1127-1130.
- Frederick E. Brot; Myron L. Bender. Use of the specificity constant of .alpha.-chymotrypsin. J. Am. Chem. Soc., 1969 , 91(25), 7187-7191.