1,7-Dioxaspiro[5.5]undecane, 98%
1,7-Dioxaspiro[5.5]undecane, 98%
1,7-Dioxaspiro[5.5]undecane, 98%
Thermo Scientific Chemicals

1,7-Dioxaspiro[5.5]undecane, 98%

CAS: 180-84-7 | C9H16O2 | 156.225 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21664.14
also known as B21664-14
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS180-84-7
IUPAC Name1,7-dioxaspiro[5.5]undecane
Molecular FormulaC9H16O2
InChI KeyGBBVHDGKDQAEOT-UHFFFAOYSA-N
SMILESC1CCC2(CCCCO2)OC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4605-1.4655 @ 20?C
FormLiquid
Suitable for use in organic farming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suitable for use in organic farming.

Solubility
Soluble in water 1.0g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Brett D Schwartz, et al. Spiroacetal biosynthesis: (+/-)-1,7-dioxaspiro[5.5]undecane in Bactrocera cacuminata and Bactrocera oleae (Olive Fruit Fly).Org. Lett.20057(6), 1173-1176.
  2. Kenji Mori and Hidenori Watanabe. Enhancement of optical purity of a product by removing unwanted diastereomers in the course of a chiral synthesis: new synthesis of the stereoisomers of 2,8-dimethyl-1,7-dioxaspiro[5.5]undecane, the pheromone of andrena wilkella.Tetrahedron.,198642(1), 295-304.