Thermo Scientific Chemicals

Trichloroacetyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: B21675.0B
1000 g, Each
Thermo Scientific Chemicals

Trichloroacetyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: B21675.0B
1000 g, Each
Quantity
Catalog number: B21675.0B
also known as B21675-0B
Price (USD)
Price: 891.00
Online price: 757.65
Your price:
Quantity
-

Chemical Identifiers

CAS
76-02-8
IUPAC Name
trichloroacetyl chloride
Molecular Formula
C2Cl4O
InChI Key
PVFOMCVHYWHZJE-UHFFFAOYSA-N
SMILES
ClC(=O)C(Cl)(Cl)Cl
Identification (FTIR)
Conforms (non-U.S. specification)
Form
Liquid
Comment
Specification differs for U.S. and non-U.S. material where indicated
Appearance (Color)
Clear colorless to pale yellow or pink
Assay (GC)
≥96.0%

Description

Trichloroacetyl chloride is used in the preparation of dihydro-1H-benzindoles. It is also used in the synthesis of 3-alkylbenzoxazolones It is used in the manufacture of pharmaceuticals and plant protection compounds. It is used for manufacturing the esters and anhydrides of trichloroacetic acid. It is used in the manufacture of pharmaceuticals and organic synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetyl chloride is used in the preparation of dihydro-1H-benzindoles. It is also used in the synthesis of 3-alkylbenzoxazolones It is used in the manufacture of pharmaceuticals and plant protection compounds. It is used for manufacturing the esters and anhydrides of trichloroacetic acid. It is used in the manufacture of pharmaceuticals and organic synthesis intermediate.

Solubility
Reacts violently with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, alcohols, amines and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text