2-Chloroethyl phenyl sulfone, 98%
2-Chloroethyl phenyl sulfone, 98%
2-Chloroethyl phenyl sulfone, 98%
Thermo Scientific Chemicals

2-Chloroethyl phenyl sulfone, 98%

CAS: 938-09-0 | C8H9ClO2S | 204.668 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B21690.14
also known as B21690-14
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS938-09-0
IUPAC Name(2-chloroethanesulfonyl)benzene
Molecular FormulaC8H9ClO2S
InChI KeyNUJGORANFDSMOL-UHFFFAOYSA-N
SMILESClCCS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)50.0-57.0?C
Assay (GC)≥97.5%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only