(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride, 97%
(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride, 97%
(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride, 97%
Thermo Scientific Chemicals

(4-Methoxycarbonylbenzyl)triphenylphosphonium chloride, 97%

CAS: 1253-47-0 | C27H24ClO2P | 446.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21729.03
also known as B21729-03
Price (USD)
30.65
Online Exclusive
34.20
Save 3.55 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
34.20
Save 3.55 (10%)
Each
Chemical Identifiers
CAS1253-47-0
IUPAC Name{[4-(methoxycarbonyl)phenyl]methyl}triphenylphosphanium chloride
Molecular FormulaC27H24ClO2P
InChI KeyNKKAHXNONUTMEY-UHFFFAOYSA-M
SMILES[Cl-].COC(=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or needles or powder or crystalline powder
Water Content (Karl Fischer Titration)< 3.0%
Appearance (Color)White to pale blue or pale grey
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only