2-Bromobutyric acid, 98%
2-Bromobutyric acid, 98%
2-Bromobutyric acid, 98%
Thermo Scientific Chemicals

2-Bromobutyric acid, 98%

CAS: 80-58-0 | C4H7BrO2 | 167.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B21743.0B
also known as B21743-0B
Price (USD)
383.65
Online Exclusive
426.00
Save 42.35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
383.65
Online Exclusive
426.00
Save 42.35 (10%)
Each
Chemical Identifiers
CAS80-58-0
IUPAC Name2-bromobutanoic acid
Molecular FormulaC4H7BrO2
InChI KeyYAQLSKVCTLCIIE-UHFFFAOYNA-N
SMILESCCC(Br)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Refractive Index1.4715-1.4755 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only